SY3H3D6829EF Synthonix Corporation

(R)-1-(o-tolyl)Butan-1-amine

  • MDL number No MDL found
  • Cas Number : 698378-41-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54978.html

Safety Information

MSDS https://synthonix.com/54978.html
Availability details for
Quantity
  • Availability
  • Add to Cart