SY3H3D6829F7 Synthonix Corporation

3-(Trifluoromethyl)quinoline-4-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 588702-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55169.html

Safety Information

MSDS https://synthonix.com/55169.html
Availability details for
Quantity
  • Availability
  • Add to Cart