SY3H3D6829F9 Synthonix Corporation

2-(4-(Trifluoromethyl)phenyl)pyrazine

  • MDL number No MDL found
  • Cas Number : 380626-88-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/55180.html

Safety Information

MSDS https://synthonix.com/55180.html
Availability details for
Quantity
  • Availability
  • Add to Cart