SY3H3D6829FA Synthonix Corporation

1-(2-(Trifluoromethyl)phenyl)-1H-pyrazole-4-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 1260855-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/55185.html

Safety Information

MSDS https://synthonix.com/55185.html
Availability details for
Quantity
  • Availability
  • Add to Cart