SY3H3D682A09 Synthonix Corporation

2-(3-(Trifluoromethyl)phenyl)pyrrolidine hydrochloride

  • Cas Number : 1197237-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55448.html

Safety Information

MSDS https://synthonix.com/55448.html
Availability details for
Quantity
  • Availability
  • Add to Cart