SY3H3D682A4F Synthonix Corporation

(S)-3-(p-Tolyl)morpholine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1391510-98-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/55492.html

Safety Information

MSDS https://synthonix.com/55492.html
Availability details for
Quantity
  • Availability
  • Add to Cart