SY3H3D682A54 Synthonix Corporation

2-(m-Tolyl)pyridine

  • MDL number No MDL found
  • Cas Number : 4373-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55630.html

Safety Information

MSDS https://synthonix.com/55630.html
Availability details for
Quantity
  • Availability
  • Add to Cart