SY3H3D682A55 Synthonix Corporation

2-(o-tolyl)pyridine

  • Cas Number : 10273-89-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55631.html

Safety Information

MSDS https://synthonix.com/55631.html
Availability details for
Quantity
  • Availability
  • Add to Cart