SY3H3D682A58 Synthonix Corporation

6-(p-Tolyl)pyridin-3-amine

  • MDL number No MDL found
  • Cas Number : 170850-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/55699.html

Safety Information

MSDS https://synthonix.com/55699.html
Availability details for
Quantity
  • Availability
  • Add to Cart