SY3H3D682A5D Synthonix Corporation

(R)-2,3,4,9-Tetrahydro-1H-carbazol-3-amine

  • MDL number No MDL found
  • Cas Number : 116650-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/55856.html

Safety Information

MSDS https://synthonix.com/55856.html
Availability details for
Quantity
  • Availability
  • Add to Cart