SY3H3D682A5F Synthonix Corporation

N-(4-(Trifluoromethyl)phenyl)pivalamide

  • MDL number No MDL found
  • Cas Number : 25617-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/55910.html

Safety Information

MSDS https://synthonix.com/55910.html
Availability details for
Quantity
  • Availability
  • Add to Cart