SY3H3D682A62 Synthonix Corporation

4,5,6-Trimethoxy-2,3-dihydro-1H-inden-1-one

  • MDL number No MDL found
  • Cas Number : 16718-42-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/55944.html

Safety Information

MSDS https://synthonix.com/55944.html
Availability details for
Quantity
  • Availability
  • Add to Cart