SY3H3D682A6C Synthonix Corporation

3',2,2-Trimethylpropiophenone

  • MDL number No MDL found
  • Cas Number : 50390-49-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56078.html

Safety Information

MSDS https://synthonix.com/56078.html
Availability details for
Quantity
  • Availability
  • Add to Cart