SY3H3D682A72 Synthonix Corporation

4,7,10,13-Tetraoxahexadecane-1,16-dinitrile

  • MDL number No MDL found
  • Cas Number : 57741-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/56257.html

Safety Information

MSDS https://synthonix.com/56257.html
Availability details for
Quantity
  • Availability
  • Add to Cart