SY3H3D682A73 Synthonix Corporation

Trimethyl 3,3',3''-phosphinetriyltripropanoate

  • MDL number No MDL found
  • Cas Number : 29269-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56296.html

Safety Information

MSDS https://synthonix.com/56296.html
Availability details for
Quantity
  • Availability
  • Add to Cart