SY3H3D682A76 Synthonix Corporation

4,4'-(1,1,3,3-Tetramethyldisiloxane-1,3-diyl)dibutanoic acid

  • Cas Number : 3353-68-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56352.html

Safety Information

MSDS https://synthonix.com/56352.html
Availability details for
Quantity
  • Availability
  • Add to Cart