SY3H3D682A77 Synthonix Corporation

1,3,5-Tripropyl-1,3,5-triazinane

  • MDL number No MDL found
  • Cas Number : 13036-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56353.html

Safety Information

MSDS https://synthonix.com/56353.html
Availability details for
Quantity
  • Availability
  • Add to Cart