SY3H3D682A7C Synthonix Corporation

1,1,1-Trifluoro-6-phenylhex-5-ene-2,4-dione

  • Cas Number : 18931-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56538.html

Safety Information

MSDS https://synthonix.com/56538.html
Availability details for
Quantity
  • Availability
  • Add to Cart