SY3H3D682A85 Synthonix Corporation

4-(4-(Trifluoromethoxy)phenoxy)piperidine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1501963-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/56590.html

Safety Information

MSDS https://synthonix.com/56590.html
Availability details for
Quantity
  • Availability
  • Add to Cart