SY3H3D682A88 Synthonix Corporation

(3S,4R)-4-(o-tolyl)Pyrrolidine-3-carboxylic acid hydrochloride

  • MDL number No MDL found
  • Cas Number : 1049727-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/56600.html

Safety Information

MSDS https://synthonix.com/56600.html
Availability details for
Quantity
  • Availability
  • Add to Cart