SY3H3D682A8D Synthonix Corporation

6-(Trifluoromethyl)-2,3,4,9-tetrahydro-1H-carbazole

  • Cas Number : 2805-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/56763.html

Safety Information

MSDS https://synthonix.com/56763.html
Availability details for
Quantity
  • Availability
  • Add to Cart