SY3H3D682A8E Synthonix Corporation

1-(3-(Trifluoromethyl)phenyl)propan-1-amine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1864062-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54130.html

Safety Information

MSDS https://synthonix.com/54130.html
Availability details for
Quantity
  • Availability
  • Add to Cart