SY3H3D682A8F Synthonix Corporation

1,2,3,4-Tetrahydrobenzo[h]quinolin-3-ol

  • MDL number No MDL found
  • Cas Number : 5423-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56815.html

Safety Information

MSDS https://synthonix.com/56815.html
Availability details for
Quantity
  • Availability
  • Add to Cart