SY3H3D682A91 Synthonix Corporation

2,3,4,9-Tetrahydro-1H-carbazole-2-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 37964-14-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/56822.html

Safety Information

MSDS https://synthonix.com/56822.html
Availability details for
Quantity
  • Availability
  • Add to Cart