SY3H3D682A94 Synthonix Corporation

1,3,3-Trimethyl-2-(formylmethylene)indoline

  • MDL number MFCD00071813
  • Cas Number : 84-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56904.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56904.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart