SY3H3D682AEE Synthonix Corporation

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(2,2,2-trifluoroethoxy)pyridine

  • Cas Number : 1415748-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57036.html

Safety Information

MSDS https://synthonix.com/57036.html
Availability details for
Quantity
  • Availability
  • Add to Cart