SY3H3D682AF7 Synthonix Corporation

1-(p-Tolyl)hexan-1-one

  • MDL number No MDL found
  • Cas Number : 1669-33-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57110.html

Safety Information

MSDS https://synthonix.com/57110.html
Availability details for
Quantity
  • Availability
  • Add to Cart