SY3H3D682AF8 Synthonix Corporation

2',2,2,5'-Tetramethylpropiophenone

  • MDL number No MDL found
  • Cas Number : 66390-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57112.html

Safety Information

MSDS https://synthonix.com/57112.html
Availability details for
Quantity
  • Availability
  • Add to Cart