SY3H3D682AFA Synthonix Corporation

(S)-2-(3,4,5-Trimethoxyphenyl)butanoic acid

  • MDL number No MDL found
  • Cas Number : 195202-08-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/57134.html

Safety Information

MSDS https://synthonix.com/57134.html
Availability details for
Quantity
  • Availability
  • Add to Cart