SY3H3D682B03 Synthonix Corporation

4,4,5,5-Tetramethyl-2-(3-(methylsulfonyl)phenyl)-1,3,2-dioxaborolane

  • MDL number No MDL found
  • Cas Number : 1001185-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57187.html

Safety Information

MSDS https://synthonix.com/57187.html
Availability details for
Quantity
  • Availability
  • Add to Cart