SY3H3D682B04 Synthonix Corporation

(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanamine

  • MDL number No MDL found
  • Cas Number : 138500-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/57208.html

Safety Information

MSDS https://synthonix.com/57208.html
Availability details for
Quantity
  • Availability
  • Add to Cart