SY3H3D682B07 Synthonix Corporation

Tridecane-1,13-diol

  • Cas Number : 13362-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57290.html

Safety Information

MSDS https://synthonix.com/57290.html
Availability details for
Quantity
  • Availability
  • Add to Cart