SY3H3D682B09 Synthonix Corporation

6-(4-(Trifluoromethyl)phenyl)picolinic acid

  • MDL number No MDL found
  • Cas Number : 924817-68-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57317.html

Safety Information

MSDS https://synthonix.com/57317.html
Availability details for
Quantity
  • Availability
  • Add to Cart