SY3H3D682B0A Synthonix Corporation

5-(4-Trifluoromethoxyphenyl)-nicotinic acid

  • MDL number No MDL found
  • Cas Number : 898796-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57318.html

Safety Information

MSDS https://synthonix.com/57318.html
Availability details for
Quantity
  • Availability
  • Add to Cart