SY3H3D682B0B Synthonix Corporation

6-(3-(Trifluoromethyl)phenoxy)picolinic acid

  • MDL number No MDL found
  • Cas Number : 137640-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57319.html

Safety Information

MSDS https://synthonix.com/57319.html
Availability details for
Quantity
  • Availability
  • Add to Cart