SY3H3D682B0D Synthonix Corporation

2,2,2-Trifluoroacetic acid compound with 2-(2-isobutyramido-6-oxo-1H-purin-9(6H)-yl)acetic acid (1:1)

  • MDL number No MDL found
  • Cas Number : 1638487-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57392.html

Safety Information

MSDS https://synthonix.com/57392.html
Availability details for
Quantity
  • Availability
  • Add to Cart