SY3H3D682B0E Synthonix Corporation

Tributylamine (dichloromethylene)bis(phosphonate)

  • Cas Number : 163706-61-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57457.html

Safety Information

MSDS https://synthonix.com/57457.html
Availability details for
Quantity
  • Availability
  • Add to Cart