SY3H3D682B15 Synthonix Corporation

2-(4-(Trifluoromethoxy)phenyl)-1,3,8-triazaspiro[4.5]dec-1-en-4-one

  • MDL number No MDL found
  • Cas Number : 1253924-71-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57641.html

Safety Information

MSDS https://synthonix.com/57641.html
Availability details for
Quantity
  • Availability
  • Add to Cart