SY3H3D682B16 Synthonix Corporation

N-(2,3,4,9-Tetrahydro-1H-carbazol-3-yl)acetamide

  • MDL number No MDL found
  • Cas Number : 60480-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57704.html

Safety Information

MSDS https://synthonix.com/57704.html
Availability details for
Quantity
  • Availability
  • Add to Cart