SY3H3D682B18 Synthonix Corporation

2,2,2-Trifluoro-1-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethanone

  • MDL number No MDL found
  • Cas Number : 631909-42-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57723.html

Safety Information

MSDS https://synthonix.com/57723.html
Availability details for
Quantity
  • Availability
  • Add to Cart