SY3H3D682B19 Synthonix Corporation

7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)quinazoline

  • Cas Number : 1375108-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57725.html

Safety Information

MSDS https://synthonix.com/57725.html
Availability details for
Quantity
  • Availability
  • Add to Cart