SY3H3D682B1B Synthonix Corporation

4-((4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)methyl)benzonitrile

  • Cas Number : 475250-43-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57787.html

Safety Information

MSDS https://synthonix.com/57787.html
Availability details for
Quantity
  • Availability
  • Add to Cart