SY3H3D682B20 Synthonix Corporation

4,4,5,5-Tetramethyl-2-(3-((2,2,2-trifluoroethyl)thio)phenyl)-1,3,2-dioxaborolane

  • MDL number No MDL found
  • Cas Number : 1026797-07-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57810.html

Safety Information

MSDS https://synthonix.com/57810.html
Availability details for
Quantity
  • Availability
  • Add to Cart