SY3H3D682B25 Synthonix Corporation

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzeneethanol

  • MDL number No MDL found
  • Cas Number : 651030-55-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57935.html

Safety Information

MSDS https://synthonix.com/57935.html
Availability details for
Quantity
  • Availability
  • Add to Cart