SY3H3D682B28 Synthonix Corporation

N1,N1,N6,N6-Tetrakis(2-hydroxyethyl)adipamide

  • MDL number No MDL found
  • Cas Number : 6334-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58007.html

Safety Information

MSDS https://synthonix.com/58007.html
Availability details for
Quantity
  • Availability
  • Add to Cart