SY3H3D682B77 Synthonix Corporation

2-(p-Tolylethynyl)aniline

  • MDL number No MDL found
  • Cas Number : 124643-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/58174.html

Safety Information

MSDS https://synthonix.com/58174.html
Availability details for
Quantity
  • Availability
  • Add to Cart