SY3H3D682B7C Synthonix Corporation

1-(5,6,7,8-Tetrahydronaphthalen-2-yl)pentan-1-one

  • Cas Number : 101498-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58381.html

Safety Information

MSDS https://synthonix.com/58381.html
Availability details for
Quantity
  • Availability
  • Add to Cart