SY3H3D682B81 Synthonix Corporation

1,1,2-Tris(tert-butoxycarbonyl)hydrazine

  • Cas Number : 185456-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58505.html

Safety Information

MSDS https://synthonix.com/58505.html
Availability details for
Quantity
  • Availability
  • Add to Cart