SY3H3D682B83 Synthonix Corporation

4'-(Trifluoromethoxy)-5-(trifluoromethyl)-[1,1'-biphenyl]-3-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 1261503-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58522.html

Safety Information

MSDS https://synthonix.com/58522.html
Availability details for
Quantity
  • Availability
  • Add to Cart