SY3H3D682B84 Synthonix Corporation

3-(Trifluoromethyl)dibenzo[b,e]oxepin-11(6H)-one

  • Cas Number : 4504-94-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/58526.html

Safety Information

MSDS https://synthonix.com/58526.html
Availability details for
Quantity
  • Availability
  • Add to Cart